1,4-Diiodo-2,5-dimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America

D5590-1G D5590-5G
TCD5590-1GEA 63 USD
TCD5590-1G TCD5590-5G
1,4-Diiodo-2,5-dimethoxybenzene ≥98.0% (by GC)
1,4-Diiodo-2,5-dimethoxybenzene
Formula: C₈H₈I₂O₂
MDL Number: MFCD00461369
CAS Number: 51560-21-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR