8-Fluoroquinoline ≥97.0% (by GC)

Supplier: TCI America
Warning

F1185-1G F1185-5G
TCF1185-1GEA 37 USD
TCF1185-1G TCF1185-5G
8-Fluoroquinoline ≥97.0% (by GC)
8-Fluoroquinoline
Formula: C₉H₆FN
MW: 147.1500 g/mol
Boiling Pt: 148 °C
MDL Number: MFCD01685514
CAS Number: 394-68-3
New Product

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR