Diallyl maleate ≥97.0% (by GC) stabilised

Supplier: TCI America
Danger

Synonyms: Diallyl Maleate

Stabilization: Stabilised with hydroquinone (HQ)

D5508-25G D5508-500G
TCD5508-25GEA 21 USD
TCD5508-25G TCD5508-500G
Diallyl maleate ≥97.0% (by GC) stabilised
Diallyl maleate
Formula: C₁₀H₁₂O₄
MW: 196.21 g/mol
Boiling Pt: 154 °C
Melting Pt: –47 °C
Flash Pt: 130 °C
MDL Number: MFCD00010763
CAS Number: 999-21-3
EINECS: 213-658-0
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR