4-(Morpholino)phenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine

T3705-1G T3705-5G
TCT3705-1GEA 124 USD
TCT3705-1G TCT3705-5G
4-(Morpholino)phenylboronic acid pinacol ester ≥98.0% (by GC)
4-(Morpholino)phenylboronic acid pinacol ester
Formula: C₁₆H₂₄BNO₃
MW: 289.18 g/mol
MDL Number: MFCD04112544
CAS Number: 568577-88-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR