3-Fluoro-2-methoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (5-Fluoro-6-methoxypyridin-3-yl)boronic acid, 3-Fluoro-2-methoxypyridine-5-boronic acid

F1237-1G F1237-200MG
TCF1237-1GEA 321 USD
TCF1237-1G TCF1237-200MG
3-Fluoro-2-methoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
3-Fluoro-2-methoxy-5-pyridineboronic acid
Formula: C₆H₇BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04112520
CAS Number: 856250-60-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR