Methyl-2-amino-4-bromobenzoate ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Amino-4-bromobenzoic acid methyll ester, Methyl 2-amino-4-bromobenzoate

M3125-25G M3125-5G
TCM3125-25GEA 187 USD
TCM3125-25G TCM3125-5G
Methyl-2-amino-4-bromobenzoate ≥97.0% (by GC)
Methyl 2-amino-4-bromobenzoate
Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD07783014
CAS Number: 135484-83-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR