Methyl-4-bromo-2-(bromomethyl)benzoate ≥97.0% (by GC)

Supplier: TCI America
Danger

M2941-1G M2941-200MG
TCM2941-1GEA 156 USD
TCM2941-1G TCM2941-200MG
Methyl-4-bromo-2-(bromomethyl)benzoate ≥97.0% (by GC)
Methyl 4-bromo-2-(bromomethyl)benzoate
Formula: C₉H₈Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04114319
CAS Number: 78471-43-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR