2-Ethylbenzoic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

E1347-25G E1347-5G
TCE1347-25GEA 194 USD
TCE1347-25G TCE1347-5G
2-Ethylbenzoic acid ≥98.0% (by GC)
2-Ethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
Boiling Pt: 259 °C
MDL Number: MFCD00045838
CAS Number: 612-19-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR