2,6-Dichloroquinoxaline ≥98.0% (by GC)

Supplier: TCI America
Warning

D5697-25G D5697-5G
TCD5697-25GEA 252 USD
TCD5697-25G TCD5697-5G
2,6-Dichloroquinoxaline ≥98.0% (by GC)
2,6-Dichloroquinoxaline
Formula: C₈H₄Cl₂N₂
MDL Number: MFCD00839695
CAS Number: 18671-97-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR