3-(4-Nitrophenyl)isoxazole ≥96.0% (by GC)

Supplier: TCI America

N1059-1G N1059-200MG
TCN1059-1GEA 270 USD
TCN1059-1G TCN1059-200MG
3-(4-Nitrophenyl)isoxazole ≥96.0% (by GC)
3-(4-Nitrophenyl)isoxazole
Formula: C₉H₆N₂O₃
MDL Number: MFCD08446390
CAS Number: 4264-05-5

Order Now

Specification Test Results

Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR