5-Fluoro-2-iodobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F1113-1G F1113-5G
TCF1113-1GEA 88 USD
TCF1113-1G TCF1113-5G
5-Fluoro-2-iodobenzoic acid ≥98.0% (by GC, titration analysis)
5-Fluoro-2-iodobenzoic acid
Formula: C₇H₄FIO₂
MW: 266.01 g/mol
MDL Number: MFCD00837316
CAS Number: 52548-63-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR