1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

E1348-1G E1348-5G
TCE1348-1GEA 68 USD
TCE1348-1G TCE1348-5G
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BN₂O₂
MDL Number: MFCD09864188
CAS Number: 847818-70-6

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR