1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(1,5-Dimethyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazole-4-boronic acid Pinacol ester

D5575-1G D5575-200MG
TCD5575-1GEA 129 USD
TCD5575-1G TCD5575-200MG
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BN₂O₂
MDL Number: MFCD09864189
CAS Number: 1036991-40-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR