Tetrakis(trimethylsiloxy)silane ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Tetrakis(trimethylsilyl) Orthosilicate, Tetrakis(trimethylsilyloxy)silane, Tetrakis(trimethylsilyl) orthosilicate

T3494-5ML T3494-25ML
TCT3494-5MLEA 48 USD
TCT3494-5ML TCT3494-25ML
Tetrakis(trimethylsiloxy)silane ≥98.0% (by GC)
Tetrakis(trimethylsiloxy)silane
Formula: C₁₂H₃₆O₄Si₅
MW: 384.85 g/mol
Boiling Pt: 220 °C
Flash Pt: 76 °C
MDL Number: MFCD00051587
CAS Number: 3555-47-3
EINECS: 222-613-4

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR