Ginkgolide B ≥93.0% (by HPLC)
Supplier: TCI America
G0553-100MG
G0553-25MG
TCG0553-100MGEA
203
USD
TCG0553-100MG
TCG0553-25MG
Ginkgolide B ≥93.0% (by HPLC)
Ginkgolide B
Formula:
C₂₀H₂₄O₁₀ Storage Temperature: Refrigerator |
MDL Number:
MFCD05662347 CAS Number: 15291-77-7 Merck Index: 14,04426 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 93.0 area% |
Elemental analysis (Carbon) | 54.00 to 59.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...