2'-Fluoro-5'-nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(2-Fluoro-5-nitrophenyl)ethanone

F1199-25G F1199-5G
TCF1199-25GEA 323 USD
TCF1199-25G TCF1199-5G
2'-Fluoro-5'-nitroacetophenone ≥98.0% (by GC)
2'-Fluoro-5'-nitroacetophenone
Formula: C₈H₆FNO₃
MW: 183.1400 g/mol
MDL Number: MFCD00277867
CAS Number: 79110-05-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR