2,2,3,4,4,4-Hexafluorobutyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: 2,2,3,4,4,4-Hexafluorobutyl Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1555-100G H1555-25G H1555-5G
TCH1555-100GEA 305 USD
TCH1555-100G TCH1555-25G TCH1555-5G
2,2,3,4,4,4-Hexafluorobutyl methacrylate ≥98.0% (by GC) stabilised
2,2,3,4,4,4-Hexafluorobutyl methacrylate
Formula: C₈H₈F₆O₂
MW: 250.14 g/mol
Flash Pt: 57 °C
MDL Number: MFCD00042311
CAS Number: 36405-47-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR