4-Ethoxy-2,3-difluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E1193-1G E1193-5G
TCE1193-1GEA 59 USD
TCE1193-1G TCE1193-5G
4-Ethoxy-2,3-difluorophenol ≥98.0% (by GC, titration analysis)
4-Ethoxy-2,3-difluorophenol
Formula: C₈H₈F₂O₂
MW: 174.14 g/mol
MDL Number: MFCD07368025
CAS Number: 126163-56-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR