4,4-Difluorocyclohexanol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5540-1G D5540-200MG
TCD5540-1GEA 135 USD
TCD5540-1G TCD5540-200MG
4,4-Difluorocyclohexanol ≥98.0% (by GC)
4,4-Difluorocyclohexanol
Formula: C₆H₁₀F₂O
Storage Temperature: Refrigerator
MDL Number: MFCD10000566
CAS Number: 22419-35-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR