4-Hydroxy-3-methylbenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H1419-1G H1419-5G
TCH1419-1GEA 53 USD
TCH1419-1G TCH1419-5G
4-Hydroxy-3-methylbenzaldehyde ≥98.0% (by GC, titration analysis)
4-Hydroxy-3-methylbenzaldehyde
Formula: C₈H₈O₂
Melting Pt: 118 °C
MDL Number: MFCD00012360
CAS Number: 15174-69-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR