3',5'-Dimethyl-2'-hydroxyacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Acetyl-4,6-dimethylphenol, 2'-Hydroxy-3',5'-dimethylacetophenone

H1569-1G H1569-5G
TCH1569-1GEA 58 USD
TCH1569-1G TCH1569-5G
3',5'-Dimethyl-2'-hydroxyacetophenone ≥98.0% (by GC)
3',5'-Dimethyl-2'-hydroxyacetophenone
Formula: C₁₀H₁₂O₂
MW: 164.2000 g/mol
Boiling Pt: 126 °C
MDL Number: MFCD01631133
CAS Number: 1198-66-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR