3-Fluoro-2-formylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Danger
|
Synonyms:
(3-Fluoro-2-formylphenyl)boronic acid, 3-Fluoro-2-formylbenzeneboronic acid
F1089-1G
F1089-5G
TCF1089-1GEA
48
USD
TCF1089-1G
TCF1089-5G
3-Fluoro-2-formylphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-2-formylphenylboronic acid
Formula:
C₇H₆BFO₃ Melting Pt: 129 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10697421 CAS Number: 871126-15-7 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...