Ethyl trimethylacetopyruvate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 5,5-dimethyl-2,4-dioxohexanoate

E1326-1G E1326-5G
TCE1326-1GEA 83 USD
TCE1326-1G TCE1326-5G
Ethyl trimethylacetopyruvate ≥98.0% (by GC)
Ethyl trimethylacetopyruvate
Formula: C₁₀H₁₆O₄
MW: 200.24
Boiling Pt: 124 °C
MDL Number: MFCD00052319
CAS Number: 13395-36-3
EINECS: 236-478-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR