3-(Trifluoromethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene

T3707-25G T3707-5G
TCT3707-25GEA 218 USD
TCT3707-25G TCT3707-5G
3-(Trifluoromethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₆BF₃O₂
MW: 272.0712 g/mol
MDL Number: MFCD05863925
CAS Number: 325142-82-3

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR