5-(Ethoxycarbonyl)-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid, 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

E1209-1G E1209-200MG
TCE1209-1GEA 177 USD
TCE1209-1G TCE1209-200MG
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211.98 g/mol
MDL Number: MFCD08235094
CAS Number: 874219-60-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR