4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

E1346-1G E1346-5G
TCE1346-1GEA 71 USD
TCE1346-1G TCE1346-5G
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥96.0% (by GC, titration analysis)
4-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03453659
CAS Number: 195062-62-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR