1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5759-25G D5759-5G
TCD5759-25GEA 359 USD
TCD5759-25G TCD5759-5G
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00085040
CAS Number: 5744-59-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR