(S)-(-)-N,α-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M3047-1ML M3047-5ML
TCM3047-1MLEA 63 USD
TCM3047-1ML TCM3047-5ML
(S)-(-)-N,α-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-N,α-Dimethylbenzylamine
Formula: C₉H₁₃N
Boiling Pt: 79 °C
Flash Pt: 61 °C
MDL Number: MFCD00067113
CAS Number: 19131-99-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -73.0 to -77.0 deg (C=2,CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR