3-Methyl-1H-1,2,4-triazole ≥98.0% (by GC)

Supplier: TCI America
Warning

M3178-1G M3178-5G
TCM3178-1GEA 54 USD
TCM3178-1G TCM3178-5G
3-Methyl-1H-1,2,4-triazole ≥98.0% (by GC)
3-Methyl-1H-1,2,4-triazole
Formula: C₃H₅N₃
Boiling Pt: 265 °C
MDL Number: MFCD00233931
CAS Number: 7170-01-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 94.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR