3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-[3-(Hydroxymethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hydroxymethylphenylboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl alcohol, (3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol, 3-(Hydroxymethyl)phenylboronic acid Pinacol ester, 3-(Hydroxymethyl)benzeneboronic acid pinacol ester

T3830-1G T3830-5G
TCT3830-1GEA 28 USD
TCT3830-1G TCT3830-5G
3-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(Hydroxymethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₃
MDL Number: MFCD09266196
CAS Number: 443776-76-9

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR