2-Formyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1108-1G F1108-5G
TCF1108-1GEA 46 USD
TCF1108-1G TCF1108-5G
2-Formyl-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Formyl-4-methoxyphenylboronic acid
Formula: C₈H₉BO₄
MW: 179.9657 g/mol
Melting Pt: 161 °C
MDL Number: MFCD01319043
CAS Number: 139962-95-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR