2-Methyl-4-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M2867-1G M2867-5G
TCM2867-1GEA 147 USD
TCM2867-1G TCM2867-5G
2-Methyl-4-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
2-Methyl-4-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD08061590
CAS Number: 660867-80-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR