4-Iodo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

I1018-1G I1018-5G
TCI1018-1GEA 56 USD
TCI1018-1G TCI1018-5G
4-Iodo-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Iodo-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃IN
MDL Number: MFCD08458091
CAS Number: 101066-87-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR