1,2-Ethylenediphosphonic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

E1265-1G E1265-5G
TCE1265-1GEA 78 USD
TCE1265-1G TCE1265-5G
1,2-Ethylenediphosphonic acid ≥97.0% (by titrimetric analysis)
1,2-Ethylenediphosphonic acid
Formula: C₂H₈O₆P₂
MDL Number: MFCD00013958
CAS Number: 6145-31-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR