Monomethyl succinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M3262-100G M3262-25G
TCM3262-100GEA 220 USD
TCM3262-100G TCM3262-25G
Monomethyl succinate ≥98.0% (by GC, titration analysis)
Monomethyl succinate
Formula: C₅H₈O₄
Boiling Pt: 151 °C
MDL Number: MFCD00002788
CAS Number: 3878-55-5
EINECS: 223-408-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR