Methyl-2-naphthoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2-naphthoate

M3169-25G M3169-5G
TCM3169-25GEA 499 USD
TCM3169-25G TCM3169-5G
Methyl-2-naphthoate ≥98.0% (by GC)
Methyl 2-naphthoate
Formula: C₁₂H₁₀O₂
Boiling Pt: 290 °C
MDL Number: MFCD00017345
CAS Number: 2459-25-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR