3,4-Dibromoiodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,2-Dibromo-4-iodobenzene

D5493-1G D5493-5G
TCD5493-1GEA 51 USD
TCD5493-1G TCD5493-5G
3,4-Dibromoiodobenzene ≥98.0% (by GC)
3,4-Dibromoiodobenzene
Formula: C₆H₃Br₂I
MDL Number: MFCD09753707
CAS Number: 909072-74-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR