O-Ethylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Ethoxyamine hydrochloride
E0880-25G
E0880-5G
TCE0880-25GEA
78
USD
TCE0880-25G
TCE0880-5G
O-Ethylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
O-Ethylhydroxylamine hydrochloride
Formula:
C₂H₈ClNO |
MDL Number:
MFCD00012956 CAS Number: 3332-29-4 EINECS: 222-060-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...