2-Ethyl-3,5-dimethylpyrazine ≥98.0% (by GC) (contains 2-Ethyl-3,6-dimethylpyrazine)

Supplier: TCI America

E1424-100G E1424-25G
TCE1424-100GEA 220 USD
TCE1424-100G TCE1424-25G
2-Ethyl-3,5-dimethylpyrazine ≥98.0% (by GC) (contains 2-Ethyl-3,6-dimethylpyrazine)
2-Ethyl-3,5-dimethylpyrazine
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Flash Pt: 69 °C
MDL Number: MFCD00047392
CAS Number: 13925-07-0
EINECS: 248-182-2

Order Now

Specification Test Results

Appearance White to Orange to Green clear liquid
Purity (GC) min. 98.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR