Di-tert-butyl phosphonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Di-tert-butyl phosphite

D5814-1G D5814-5G
TCD5814-1GEA 26 USD
TCD5814-1G TCD5814-5G
Di-tert-butyl phosphonate ≥98.0% (by titrimetric analysis)
Di-tert-butyl phosphonate
Formula: C₈H₁₉O₃P
MW: 194.21
Boiling Pt: 66 °C
Flash Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00995019
CAS Number: 13086-84-5
EINECS: 235-996-8

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (Neutralization titration) min. 98.0 %
Purity (Neutralization back titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR