2-Phenyl-1,3-dithiane ≥98.0% (by GC)

Supplier: TCI America

P2660-25G P2660-5G
TCP2660-25GEA 161 USD
TCP2660-25G TCP2660-5G
2-Phenyl-1,3-dithiane ≥98.0% (by GC)
2-Phenyl-1,3-dithiane
Formula: C₁₀H₁₂S₂
MDL Number: MFCD00006656
CAS Number: 5425-44-5
EINECS: 226-568-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR