N-(2-Pyridyl)thiourea ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

P2272-25G P2272-5G
TCP2272-25GEA 422 USD
TCP2272-25G TCP2272-5G
N-(2-Pyridyl)thiourea ≥98.0% (by HPLC, titration analysis)
N-(2-Pyridyl)thiourea
Formula: C₆H₇N₃S
MDL Number: MFCD00041227
CAS Number: 14294-11-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR