(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,3-Dibromopropionic acid methyll ester, Methyl 2,3-Dibromopropionate, Methyl 2,3-dibromopropanoate

M3182-25G M3182-5G
TCM3182-25GEA 110 USD
TCM3182-25G TCM3182-5G
(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)
(±)-Methyl-2,3-dibromopropionate
Formula: C₄H₆Br₂O₂
MW: 245.9 g/mol
Boiling Pt: 204 °C
MDL Number: MFCD00017882
CAS Number: 1729-67-5
EINECS: 217-044-3

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR