Diethyl phenylphosphonite ≥96.0% (by GC)

Supplier: TCI America
Danger

D5593-1G D5593-5G
TCD5593-1GEA 34 USD
TCD5593-1G TCD5593-5G
Diethyl phenylphosphonite ≥96.0% (by GC)
Diethyl phenylphosphonite
Formula: C₁₀H₁₅O₂P
Boiling Pt: 235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009086
CAS Number: 1638-86-4
EINECS: 216-676-7
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR