Methyl-4-bromo-3-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 4-bromo-3-fluorobenzoate

M3083-25G M3083-5G
TCM3083-25GEA 362 USD
TCM3083-25G TCM3083-5G
Methyl-4-bromo-3-fluorobenzoate ≥98.0% (by GC)
Methyl-4-bromo-3-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD08436044
CAS Number: 849758-12-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR