4-Fluoro-3-nitrobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

F1040-5G F1040-25G
TCF1040-5GEA 46 USD
TCF1040-5G TCF1040-25G
4-Fluoro-3-nitrobenzonitrile ≥98.0% (by GC)
4-Fluoro-3-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MDL Number: MFCD01632197
CAS Number: 1009-35-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR