4-Fluorophenethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

F1256-1G F1256-5G
TCF1256-1GEA 56 USD
TCF1256-1G TCF1256-5G
4-Fluorophenethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluorophenethylamine hydrochloride
Formula: C₈H₁₁ClFN
Melting Pt: 208 °C
MDL Number: MFCD00008190
CAS Number: 459-19-8
EINECS: 207-285-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR