SKF-96365 hydrochloride ≥98% (by HPLC)
Supplier: TCI America
Synonyms:
1-(2-(4-Methoxyphenyl)-2-(3-(4-methoxyphenyl)propoxy)ethyl)-1H-imidazole hydrochloride, 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole hydrochloride, SKF-96365
K0066-10MG
K0066-50MG
TCK0066-10MGEA
231
USD
TCK0066-10MG
TCK0066-50MG
SKF-96365 hydrochloride ≥98% (by HPLC)
SKF-96365 hydrochloride
Formula:
C₂₂H₂₆N₂O₃·HCl MW: 402.9 g/mol Melting Pt: 119 °C |
MDL Number:
MFCD00236407 CAS Number: 130495-35-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 6.60 to 7.30 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...