2-Isopropoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

I1099-1G I1099-200MG
TCI1099-1GEA 105 USD
TCI1099-1G TCI1099-200MG
2-Isopropoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Isopropoxy-5-pyridineboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₃
MDL Number: MFCD07368871
CAS Number: 871839-91-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR