4-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

T3710-1G T3710-5G
TCT3710-1GEA 28 USD
TCT3710-1G TCT3710-5G
4-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC)
4-Nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR